Learn More
Invitrogen™ Caspase 7 p11 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, HeLa, HepG2, Jurakt, Jurakt (10 μM etoposide treatment for 24 hr). Predicted reactivity: Mouse (91%), Rat (91%), Zebrafish (83%), Xenopus laevis (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Caspase 7 p11 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.93 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | CASP7 |
| Gene Accession No. | P55210 |
| Gene Alias | AI314680; Apoptotic protease Mch-3; Casp7; CASP-7; caspase 7; caspase 7, apoptosis-related cysteine peptidase; caspase 7, apoptosis-related cysteine protease; caspase-7; Caspase-7 subunit p11; Caspase-7 subunit p20; CMH-1; cysteine protease LICE2; ICE-IAP3; ICE-LAP3; ICE-like apoptotic protease 3; LICE2; mCASP-7; MCH3 |
| Gene Symbols | CASP7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.