Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-8 Antibody (1G12), DyLight 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280097DL594
This item is not returnable.
View return policy
Description
Caspase-8 Monoclonal antibody specifically detects Caspase-8 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
Caspase-8 | |
Monoclonal | |
DyLight 594 | |
AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
Recombinant mouse caspase-8 (p20 subunit). | |
0.1 mL | |
Apoptosis, Apoptosis Detections, Cancer, Caspases, Cell Biology, Necroptosis, Phospho Specific, Ubiquitin Proteasome Pathway | |
841 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
1G12 | |
50mM Sodium Borate | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction