Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 61 publication
Supplier: Novus Biologicals NB10056116
Description
Caspase-8 Polyclonal specifically detects Caspase-8 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Caspase-8 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Simple Western reported by internal validation, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen 1:10-1:500 | |
| Q14790 | |
| CASP8 | |
| Recombinant catalytically active human Caspase-8 protein. | |
| Unpurified | |
| RUO | |
| Primary | |
| Detects Caspase-8 and cleavage products. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
| Rabbit | |
| 55.4 kDa | |
| 0.05 mL | |
| Apoptosis, Apoptosis Detections, Cancer, Caspases, Cell Biology, Phospho Specific, Ubiquitin Proteasome Pathway | |
| 841 | |
| Human, Mouse, Rat, Gerbil | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction