Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-8 Antibody (90A992), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056527C
Description
Caspase-8 Monoclonal specifically detects Caspase-8 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Caspase-8 | |
Monoclonal | |
DyLight 650 | |
Q14790 | |
CASP8 | |
The immunogen for this product falls between amino acids 360-385 of the human Caspase 8 protein. | |
0.1 mL | |
Apoptosis, Cancer, Caspases, Cell Biology, Ubiquitin Proteasome Pathway | |
841 | |
Human, Primate | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
90A992 | |
Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin | |
AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This can be used for detection of the pro-form of Caspase-8. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction