Learn More
Invitrogen™ Caspase 8 (Cleaved Asp374) Recombinant Rabbit Monoclonal Antibody (RM442)

Description
This product targets amino acids 375-479 of Caspase 8.
Specifications
Specifications
| Antigen | Caspase 8 (Cleaved Asp374) |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | RM442 |
| Concentration | 50 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.09% sodium azide |
| Gene | CASP8 |
| Gene Accession No. | Q14790 |
| Gene Alias | ALPS2B; Apoptotic cysteine protease; apoptotic protease Mch-5; CAP4; CASP8; Casp-8; caspase 8; caspase 8, apoptosis-related cysteine peptidase; caspase 8, apoptosis-related cysteine protease; caspase-8; Caspase-8 subunit p10; Caspase-8 subunit p18; FADD-homologous ICE/ced-3-like protease; FADD-like ICE; FLICE; ICE-like apoptotic protease 5; MACH; MACH-alpha-1/2/3 protein; MACH-beta-1/2/3/4 protein; MCH5; MORT1-associated ced-3 homolog |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.