Specifications
| Quantity | 100 Tests |
| Content And Storage | 100 mL Cell Lysis Buffer, 4 x 2 mL 2X Reaction Buffer, 0.5 mL IETD-AFC (1 mM), & 0.4 mL DTT (1 M). Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |