Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caspase 8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120116
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120116 Supplier Invitrogen™ Supplier No. PA5120116
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: THP-1, Mouse liver. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: QSCLYAIAEE LGSEDLAALK FLCLDYIPHK KQETIEDAQK LFLRLREKGM LEEGNLSFLK ELLFHISRWD LLVNFLDCNR EEMVRELRDP DNAQISPYRV MLFKLSEEVS ELELRSFKFL LNNEIPKCKL EDDLSLLEIF VEMEKRTMLA ENNLETLKSI CDQVNKSLLG KIEDYERSST ERRMSLEGRE ELPPSVLDEM SLKMAELCDS PREQDSESRT SDKVYQMKNK PRGYCLIINN HDFSKAREDI TQLRKMKDRK GTDCDKEALS KTFKELHFEI VSYDDCTANE IHEILEGYQS ADHKNKDCFI CCILSHGDKG VVYGTDGKEA SIYDLTSYFT GSKCPSLSGK PKIFFIQACQ GSNFQKGVPD EAGFEQQNHT LEVDSSSHKN YIPDEADFLL GMATVKNCVS YRDPVNGTWY IQSLCQSLRE RCPQGDDILS ILTGVNYDVS NKDDRRNKGK QMPQPTFTLR KKLFFPP.

Caspase 8 (CASP8) is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases play a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes composed of a pro-domain, a large protease subunit, and a small protease subunit. Activation of caspases requires proteolytic processing at conserved internal aspartic residues to generate a heterodimeric enzyme consisting of the large and small subunits. Caspase 8 is involved in the programmed cell death induced by Fas and various apoptotic stimuli. The N-terminal FADD-like death effector domain of Caspase 8 suggests that it may interact with Fas-interacting protein FADD. Caspase 8 was detected in the insoluble fraction of the affected brain region from Huntington disease patients but not in those from normal controls, which implicated the role in neurodegenerative diseases. Caspase 8 binds to the death effector domain (DED) of FADD through an analogous DED domain present in tandem in the pro-form of the Caspase 8 protein. Activated Caspase 8 then activates other downstream caspases including Caspase 9, thereby committing the cell to undergo apoptosis. In addition, Caspase 8 also reacts with Jurkat cells and Tonsil. Overexpression of Caspase 8 induces apoptosis, which can be blocked by inhibitors specific for the ICE family. Many alternatively spliced transcript variants encoding different isoforms have been described for Caspase 8, however, not all variants have had their full-length sequences determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 8
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Casp8
Gene Accession No. O89110, Q14790
Gene Alias ALPS2B; apoptosis-related cysteine peptidase; apoptotic cysteine protease; Apoptotic protease Mch-5; CAP4; CASP8; CASP-8; caspase 8; caspase 8, apoptosis-related cysteine peptidase; caspase 8, apoptosis-related cysteine protease; Caspase8; caspase-8; Caspase-8 precursor; Caspase-8 subunit p10; Caspase-8 subunit p18; EC 3.4.22.61; FADD-homologous ICE/ced-3-like protease; FADD-like ICE; Fas-linked ICE-like protease; FLICE; FLJ17672; ICE8; ICE-like apoptotic protease 5; MACH; MACH-alpha-1/2/3 protein; MACH-beta-1/2/3/4 protein; MCH5; MGC78473; MORT1-associated ced-3 homolog; OTTHUMP00000163720; OTTHUMP00000165063; OTTHUMP00000206557; OTTHUMP00000206581
Gene Symbols Casp8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 4-480 of mouse Caspase 8 (NP_0339421).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12370, 841
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.