Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-8 Rat anti-Mouse, Clone: 3B10, MilliporeSigma™

Catalog No. MABC160625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1606-25 25 μg
MABC1606-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1606-25 Supplier MilliporeSigma Supplier No. MABC160625UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Caspase-8, clone 3B10 is a rat monoclonal antibody that detects Caspase-8 and has been tested for use in Flow Cytometry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase-8
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 3B10
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Casp8
Host Species Rat
Immunogen Recombinant p20 subunit of Mouse Caspase-8.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001073595
Test Specificity This rat monoclonal antibody detects Caspase-8 in murine species. It targets an epitope within the p20 subunit.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.