Learn More
Description
Specifications
Specifications
| Antigen | Caspase-9 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:200-1:500, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:200-1:500 |
| Formulation | Whole antisera with 0.02% Sodium Azide |
| Gene Alias | APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease |
| Gene Symbols | CASP9 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to amino acids 188-206 (EKLRRRFSSLHFMVEVKGD) of human caspase-9 was used as immunogen; GenBank no. gi 27802691 gb AAO21133.1 . |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
