Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-9 Antibody (8G5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Caspase-9 |
---|---|
Clone | 8G5 |
Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Flow Cytometry, ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
Caspase-9 Monoclonal antibody specifically detects Caspase-9 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
Caspase-9 | |
Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease | |
A synthesized peptide derived from Human Caspase-9 [UniProt P55211] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
8G5 | |
Flow Cytometry, ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Apoptosis, Cancer, Caspases, Cell Biology, Neurodegeneration, Neuroscience, Phospho Specific | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
842 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title