Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-9 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056121T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056121T 0.025 mL
NB10056121 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056121T Supplier Novus Biologicals Supplier No. NB10056121SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Caspase-9 Polyclonal specifically detects Caspase-9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Caspase-9
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:200-1:500, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:200-1:500
Formulation Whole antisera with 0.02% Sodium Azide
Gene Alias APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease
Gene Symbols CASP9
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 188-206 (EKLRRRFSSLHFMVEVKGD) of human caspase-9 was used as immunogen; GenBank no. gi 27802691 gb AAO21133.1 .
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 842
Test Specificity Caspase-9
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.