Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Caspase-Glo 1 Inflammasome Assay, 10ml

Catalog No. PRG9951 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG9951 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG9951 Supplier Promega Supplier No. G9951
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Caspase-Glo 1 Inflammasome Assay is a homogeneous, bioluminescent method to selectively measure the activity of caspase-1, a member of the cysteine aspartic acid-specific protease (caspase) family and an essential component of the inflammasome.

  • A Bioluminescent Method to Directly Measure Inflammasome Activity
  • Fast, simple, homogeneous assay
  • Selective and quantitative
  • Multiplex capability

Specifications

Quantity 10 mL
Content And Storage -30°C to -10°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.