Learn More
Description
- A Biochemical Assay That Measures Caspase-6 Activity
- Simple “add-mix-measure protocol amenable to automation
- Greater sensitivity than fluorescence-based assays
Specifications
Specifications
| Quantity | 10 mL |
| pH | 7.2 |
| Kit Contents | 1 x 10mL Caspase-Glo 6 Buffer, 1 x 100μL Z-VEID-Glo substrate, 1 x 1 each Luciferin Detection Reagent |
| Test Time | 30 min. |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | Caspase-6 Enzyme Studies, Caspase-6 Inhibitor Screening |
| Form | Fluid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.