Learn More
Description
- Advantages: simplified method, greater sensitivity provide faster results, increased accuracy
- Batch-processing capability
- Includes: Caspase-Glo 6 Buffer, Z-VEID-Glo™ Substrate, Luciferin Detection Reagent
- Ideal for biochemical screens for Caspase -2 inhibitors
Specifications
Specifications
| Quantity | 50 mL |
| pH | 7.2 |
| Kit Contents | 1 x 50mL Caspase-Glo 6 Buffer, 1 x 500μL Z-VEID-Glo substrate, 1 x 1 each Luciferin Detection Reagent |
| Test Time | 30 min. |
| Content And Storage | -20°C |
| For Use With (Application) | Caspase-6 Enzyme Studies, Caspase-6 Inhibitor Screening |
| Form | Fluid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.