Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspr1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159251
Description
Caspr1 Polyclonal specifically detects Caspr1 in Human, Goat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Caspr1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Caspr, Caspr1, CASPRcaspr1, CNTNAP, contactin associated protein 1, neurexin 4, Neurexin IV, Neurexin-4, NRXN4contactin-associated protein 1, P190 | |
Rabbit | |
Affinity purified | |
RUO | |
8506 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunocytochemistry/Immunofluorescence | |
P78357 | |
CNTNAP1 | |
Synthetic peptides corresponding to CNTNAP1(contactin associated protein 1) The peptide sequence was selected from the N terminal of CNTNAP1. Peptide sequence LQIDLMKKHRIRAVATQGSFNSWDWVTRYMLLYGDRVDSWTPFYQRGHNS. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Rabbit: 100%; Mouse: 92%; Rat: 92%. | |
Human, Goat, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction