Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspr1 Antibody (SR1234), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32178950UL
Description
Caspr1 Monoclonal antibody specifically detects Caspr1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Caspr1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR1234 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Caspr, Caspr1, CASPRcaspr1, CNTNAP, contactin associated protein 1, neurexin 4, Neurexin IV, Neurexin-4, NRXN4contactin-associated protein 1, P190 | |
A synthesized peptide derived from human Caspr1 (Uniprot #: P78357) | |
50 μg | |
Neuroscience | |
8506 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction