Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspr2 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298329B
Description
Caspr2 Polyclonal specifically detects Caspr2 in Mouse samples. It is validated for ELISA.Specifications
Caspr2 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
26047 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Biotin | |
AUTS15, CASPR2DKFZp781D1846, CDFE, Cell recognition molecule Caspr2, contactin associated protein-like 2, contactin-associated protein 2, contactin-associated protein-like 2, homolog of Drosophila neurexin IV, KIAA0868, NRXN4Caspr2, PTHSL1 | |
Produced in rabbits immunized with purified, recombinant Caspr2 (NP_001004357.2; Met 1-Ser 1262). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction