Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Caspr2 Rabbit anti-Mouse, Clone: 1, Novus Biologicals™
 
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
| Antigen | Caspr2 | 
|---|---|
| Clone | 1 | 
| Dilution | ELISA 1:5000-1:10000 | 
| Applications | ELISA | 
| Classification | Monoclonal | 
Description
Caspr2 Monoclonal specifically detects Caspr2 in Mouse samples. It is validated for ELISA.Specifications
| Caspr2 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 26047 | |
| IgG | |
| Protein A purified | 
| 1 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| AUTS15, CASPR2DKFZp781D1846, CDFE, Cell recognition molecule Caspr2, contactin associated protein-like 2, contactin-associated protein 2, contactin-associated protein-like 2, homolog of Drosophila neurexin IV, KIAA0868, NRXN4Caspr2, PTHSL1 | |
| Recombinant Mouse Caspr2 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            