Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caspr2 Recombinant Rabbit Monoclonal Antibody (2H2I5)
GREENER_CHOICE

Catalog No. PIMA537891
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537891 Supplier Invitrogen™ Supplier No. MA537891
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Mouse spinal cord, Rat brain Immunogen sequence: HLDHLDSASA DFPYNPGQGQ AIRNGVNRNS AIIGGVIAVV IFTILCTLVF LIRYMFRHKG TYHTNEAKGA ESAESADAAI MNNDPNFTET IDESKKEWLI.

This gene encodes a member of the neurexin family which functions in the vertebrate nervous system as cell adhesion molecules and receptors. This protein, like other neurexin proteins, contains epidermal growth factor repeats and laminin G domains. In addition, it includes an F5/8 type C domain, discoidin/neuropilin- and fibrinogen-like domains, thrombospondin N-terminal-like domains and a putative PDZ binding site. This protein is localized at the juxtaparanodes of myelinated axons, and mediates interactions between neurons and glia during nervous system development and is also involved in localization of potassium channels within differentiating axons. This gene encompasses almost 1.5% of chromosome 7 and is one of the largest genes in the human genome. It is directly bound and regulated by forkhead box protein P2 (FOXP2), a transcription factor related to speech and language development. This gene has been implicated in multiple neurodevelopmental disorders, including Gilles de la Tourette syndrome, schizophrenia, epilepsy, autism, ADHD and mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspr2
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 2H2I5
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CNTNAP2
Gene Accession No. Q9CPW0
Gene Alias 5430425M22Rik; AUTS15; CASPR2; CDFE; Cell recognition molecule Caspr2; Cntnap2; contactin associated protein-like 2; contactin-associated protein-like 2; DKFZp781D1846; homolog of Drosophila neurexin IV; KIAA0868; mKIAA0868; NRXN4; PTHSL1
Gene Symbols CNTNAP2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1232-1331 of human Caspr2/CNTNAP2 (Q9UHC6).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 500105, 66797
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.