Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CASS4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568383
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568383 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568383 Supplier Invitrogen™ Supplier No. PA568383
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CASS4 Polyclonal Antibody for IHC (P)

Docking protein that plays a role in tyrosine kinase-based signaling related to cell adhesion and cell spreading. Regulates PTK2/FAK1 activity, focal adhesion integrity, and cell spreading. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CASS4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CASS4
Gene Accession No. Q9NQ75
Gene Alias C20orf32; cas scaffolding protein family member 4; CAS4; CASS4; HEF1-EFS-p130Cas-like protein; HEFL; HEF-like protein; HEPL
Gene Symbols CASS4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human CASS4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57091
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.