Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CASS4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548588
Description
Docking protein that plays a role in tyrosine kinase- based signaling related to cell adhesion and cell spreading. Regulates PTK2/FAK1 activity, focal adhesion integrity, and cell spreading.
Specifications
CASS4 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9NQ75 | |
CASS4 | |
KLH conjugated synthetic peptide between 610-639 amino acids from the C-terminal region of human CASS4. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CASS4 | |
Cas scaffolding protein family member 4, HEF-like protein, HEF1-EFS-p130Cas-like protein, HEPL, CASS4, C20orf32, HEFL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57091 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction