Learn More
Castor Oil, Sulfated, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Castor Oil, Sulfated is an oil that is derived from the castor beans and is made by adding sulfuric acid to the oil. A colorless oil that has no taste or smell and boils at 313°C, it is used as a synthetic detergent in formulating lubricants, softeners, and dyeing assistants. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
CAS Max % | 68-76%, 24-32% |
Packaging | Amber Glass Bottle |
Quantity | 500 mL |
Grade | Reagent |
Physical Form | Viscous Liquid or Low Melting Solid |
Chemical Name or Material | Castor Oil |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.