Learn More
Description
Specifications
Specifications
| Antigen | CASZ1 |
| Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Formulation | Guinea Pig polyclonal antibody serum with 0.05% Sodium Azide. |
| Gene | Casz1, Cst, D4Ertd432e, Kiaa3026, Srg, Znf693 |
| Gene Accession No. | Q9CWL2 |
| Host Species | Guinea Pig |
| Immunogen | A mixture of three peptides representing two internal sequences N-terminal to the first zinc finger and a third sequence within the region between the third and fourth zinc fingers. |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.