Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CASZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569006
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569006 Supplier Invitrogen™ Supplier No. PA569006
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

The protein encoded by this gene is a zinc finger transcription factor. The encoded protein may function as a tumor suppressor, and single nucleotide polymorphisms in this gene are associated with blood pressure variation. Alternative splicing results in multiple transcript variants that encode different protein isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CASZ1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CASZ1
Gene Accession No. 0
Gene Alias 2410019P08Rik; AV096627; BC040081; CAS11; castor homolog 1, zinc finger; castor zinc finger 1; castor-related protein; Casz1; CST; D4Ertd432e; dJ734G22.1; Kiaa3026; MTMR9; Putative survival-related protein; RGD1563533; sb:eu580; SRG; survival-related; wu:fk85f10; zinc finger protein 693; Zinc finger protein castor homolog 1; zinc finger protein castor homolog 1; LOW QUALITY PROTEIN: zinc finger protein castor homolog 1; ZNF693
Gene Symbols CASZ1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human CASZ1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313713
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.