Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CASZ1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598568
Description
CASZ1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene is a zinc finger transcription factor. The encoded protein may function as a tumor suppressor, and single nucleotide polymorphisms in this gene are associated with blood pressure variation. Alternative splicing results in multiple transcript variants that encode different protein isoforms.
Specifications
CASZ1 | |
Polyclonal | |
Unconjugated | |
CASZ1 | |
2410019P08Rik; AV096627; BC040081; CAS11; castor homolog 1, zinc finger; castor zinc finger 1; castor-related protein; Casz1; CST; D4Ertd432e; dJ734G22.1; Kiaa3026; MTMR9; Putative survival-related protein; RGD1563533; sb:eu580; SRG; survival-related; wu:fk85f10; zinc finger protein 693; Zinc finger protein castor homolog 1; zinc finger protein castor homolog 1; LOW QUALITY PROTEIN: zinc finger protein castor homolog 1; ZNF693 | |
Rabbit | |
Protein G | |
RUO | |
54897 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4.31 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q86V15 | |
CASZ1 | |
Recombinant Human Zinc finger protein castor homolog 1 protein (127-266AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction