Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASZ1 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992005
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2005 25 μL
50-199-2004 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2005 Supplier Rockland Immunochemicals Supplier No. 600401B62S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human CASZ1 protein. A BLAST analysis was used to suggest reactivity with CASZ1 proteins from human, mouse, Drosophila, chimpanzee, and macaque based on a 100% homology. Partial reactivity is expected with horse and dog CASZ1 based on a 92% homology with the immunizing sequence. Cross-reactivity with CASZ1 from other sources has not been determined.

The protein encoded by this gene is a zinc finger transcription factor. The encoded protein may function as a tumor suppressor, and single nucleotide polymorphisms in this gene are associated with blood pressure variation. Alternative splicing results in multiple transcript variants that encode different protein isoforms.

Specifications

Antigen CASZ1
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene CASZ1
Gene Accession No. Q86V15, Q9CWL2
Gene Alias 2410019P08Rik; AV096627; BC040081; BOS_15820; CAS11; castor homolog 1, zinc finger; castor zinc finger 1; Castor-related protein; Casz1; Cst; D4Ertd432e; dJ734G22.1; Kiaa3026; Putative survival-related protein; RGD1563533; sb:eu580; SRG; survival-related; wu:fk85f10; Zinc finger protein 693; Zinc finger protein castor homolog 1; ZNF693
Gene Symbols CASZ1
Host Species Rabbit
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 457931, 54897, 69743, 722677
Target Species Chimpanzee, Drosophila, Human, Mouse, Rhesus Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.