Learn More
CAT-1 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
Specifications
Specifications
| Antigen | CAT-1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | SLC7A1 |
| Gene Accession No. | P30825 |
| Gene Alias | amino acid transporter, cationic 1, ATRC1, CAT1, CAT-1, ecotropic retroviral leukemia receptor homolog, ecotropic retroviral receptor, ecotropic retrovirus receptor homolog, ERR, HCAT1, REC1L, solute carrier family 7 (cationic amino acid transporter, y+ system), member 1, Solute carrier family 7 member 1, system Y+ basic amino acid transporter |
| Gene Symbols | SLC7A1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.