Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAT Mouse anti-Canine, Human, Rat, Clone: OTI1B6, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA502496

Description
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kD in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara′s disease.Specifications
CAT | |
Monoclonal | |
0.72 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O97492, P04040, P04762 | |
CAT | |
Full length human recombit protein of human CAT produced in HEK293T cell. | |
100 μL | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1B6 | |
Unconjugated | |
CAT | |
catalase | |
Mouse | |
Affinity Chromatography | |
RUO | |
24248, 403474, 847 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction