Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cat Mouse anti-Human, Rat, Clone: 12C2DB9, Abnova™

Catalog No. 89035413 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-413 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-413 Supplier Abnova Corporation Supplier No. MAB14008

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against rat Cat.

  • High specificity
  • High reactivity

Specifications

Antigen Cat
Applications Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 12C2DB9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against rat Cat.
Dilution Immunocytochemistry (5 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Formulation In HEPES (0.02% sodium azide).
Gene Accession No. P04762
Gene Alias CS1/Cas1/Cs-1/RATCAT01/RATCATL
Gene Symbols Cat
Host Species Mouse
Immunogen Rat liver mitochondria.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24248
Target Species Human, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.