Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cat Rabbit anti-Human, Mouse, Rat, Polyclonal, Abnova™

Catalog No. 89035689 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-689 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-689 Supplier Abnova Corporation Supplier No. PAB30196

Rabbit Polyclonal Antibody

Rabbit polyclonal antibody raised against synthetic peptide of mouse Cat.

  • High specificity
  • High reactivity

Specifications

Antigen Cat
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of mouse Cat.
Dilution ELISA (0.05-0.2 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.4 (0.02% sodium azide).
Gene Accession No. P24270
Gene Alias 2210418N07/Cas-1/Cas1/Cs-1
Gene Symbols Cat
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus of mouse Cat.
Purification Method Immunoaffinity purification
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12359
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C on dry atmosphere, lyophilized antibodies are stable.
After reconstitution with deionized water, store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.