Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Cat/Rabbit Ventilator
SDP

Stoelting™ Cat/Rabbit Ventilator features a unique design that ensures accuracy, durability and quietness of operation.

$17860.00 - $20610.00

Specifications

For Use With (Application) Neuroscience
Animal Cat, Rabbit
View More Specs

Products 4
Catalog Number Mfr. No. Model Size Price Quantity & Availability  
Catalog Number Mfr. No. Model Size Price Quantity & Availability  
10-001-072
SDP
View Documents
Stoelting™
501955
Ventilator 50 mL
Each for $17,860.00
Only null left
Add to Cart
 
10-001-094
SDP
View Documents
Stoelting™
561255
Ventilator with synchronized start/stop 50 mL
Each for $20,610.00
Only null left
Add to Cart
 
10-001-104
SDP
View Documents
Stoelting™
5019510
Ventilator 100 mL
Each for $17,860.00
Only null left
Add to Cart
 
10-000-954
SDP
View Documents
Stoelting™
56125
Ventilator with synchronized start/stop 100 mL
Each for $20,610.00
Only null left
Add to Cart
 
Description

Description

  • Valve mechanism features nearly zero dead space and superb durability with no leakage. Expired air may be channeled for analysis and/or recycled, when used with anesthetic gases.
  • Digital stroke rate LED display
  • User selected pause (Model 56125) - can be paused at any point in the respiration cycle for taking measurements or for gas analysis.
  • Easily adjustable stroke volume whether the ventilator is on or off
    • For the most convenient and accurate stroke volume control available, the knob and scale remain stationary, even when the piston is in motion. 
  • Elimination of mechanical noise, reduced wear, and very low maintenance requirements
  • Cylinder/piston assemblies available
    • Cat units are supplied with one of two easily interchangeable cylinder/piston assemblies that enable a broader range of stroke volumes
Specifications

Specifications

Neuroscience
Cat, Rabbit
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.