Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAT1 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232271V
Description
CAT1 Polyclonal antibody specifically detects CAT1 in Human samples. It is validated for Western BlotSpecifications
CAT1 | |
Polyclonal | |
Western Blot | |
amino acid transporter, cationic 1, ATRC1ERR, CAT-1System Y+ basic amino acid transporter, Ecotropic retroviral leukemia receptor homolog, ecotropic retroviral receptor, Ecotropic retrovirus receptor homolog, HCAT1, high affinity cationic amino acid transporter 1, REC1LCAT1, solute carrier family 7 (cationic amino acid transporter, y+ system), member 1, Solute carrier family 7 member 1 | |
The immunogen this antibody was made to, maps to a region between residue 600 to 650 of human High Affinity Cationic Amino Acid Transporter 1 using the numbering given in entry NP_003036.1 (GeneID 6541). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
6541 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction