Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582800
Description
Immunogen sequence: YQILSSCRVP GPKQDTVSNF SKTKKPPTHI TVVHNYQFFE LDVYHSDGTP LTADQIFVQL EKIWNSSLQT NKEPVGILTS NHRNSWAKAY NTLIK.
Carnitine acetyltransferase (CRAT) is a key enzyme in the metabolic pathway in mitochondria, peroxisomes and endoplasmic reticulum. CRAT catalyzes the reversible transfer of acyl groups from an acyl-CoA thioester to carnitine and regulates the ratio of acylCoA/CoA in the subcellular compartments. Different subcellular localizations of the CRAT mRNAs are thought to result from alternative splicing of the CRAT gene suggested by the divergent sequences in the 5' region of peroxisomal and mitochondrial CRAT cDNAs and the location of an intron where the sequences diverge. The alternatively splicing of this gene results in three distinct isoforms, one of which contains an N-terminial mitochondrial transit peptide, and has been shown to be located in mitochondria.
Specifications
| CAT1 | |
| Polyclonal | |
| Unconjugated | |
| CRAT | |
| amino acid transporter, cationic 1; ATRC1; AW107812; CARAT; Carnitine acetylase; carnitine acetyltransferase; Carnitine O-acetyltransferase; CAT; CAT1; CAT-1; Crat; Ecotropic retroviral leukemia receptor homolog; ecotropic retroviral receptor; ecotropic retrovirus receptor homolog; ERR; HCAT1; High affinity cationic amino acid transporter 1; REC1L; SLC7A1; solute carrier family 7 (cationic amino acid transporter, y+ system), member 1; solute carrier family 7 member 1; System Y+ basic amino acid transporter | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1384 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P43155 | |
| CRAT | |
| Recombinant protein corresponding to Human CRAT. Recombinant protein control fragment (Product #RP-92891). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction