Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109589
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109589 Supplier Invitrogen™ Supplier No. PA5109589
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLRFNITPEI KSDIEKAKQN LSIMIQDLDI TVMVFHHFGK DFPKSEKLSP DAFIQMALQL AYYRIYGQAC ATYESASLRM FHLGRTDTIR SASMDSLTFV KAMDDSSVTE HQKVELLRKA VQAHRGYTDR AIRGEAFDRH LLGLKLQAIE DLVSMPDIFM DTSYAIAMHF HLSTSQVPAK TDCVMFFGPV VPDGYGVCYN PMEAHINFSL SAYNSCAETN AARLAHYLEK ALLDMRALLQ SHPRAKL.

Carnitine acetyltransferase (CRAT) is a key enzyme in the metabolic pathway in mitochondria, peroxisomes and endoplasmic reticulum. CRAT catalyzes the reversible transfer of acyl groups from an acyl-CoA thioester to carnitine and regulates the ratio of acylCoA/CoA in the subcellular compartments. Different subcellular localizations of the CRAT mRNAs are thought to result from alternative splicing of the CRAT gene suggested by the divergent sequences in the 5' region of peroxisomal and mitochondrial CRAT cDNAs and the location of an intron where the sequences diverge. The alternatively splicing of this gene results in three distinct isoforms, one of which contains an N-terminial mitochondrial transit peptide, and has been shown to be located in mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAT1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CRAT
Gene Accession No. P43155, P47934, Q704S8
Gene Alias amino acid transporter, cationic 1; ATRC1; AW107812; CARAT; Carnitine acetylase; carnitine acetyltransferase; Carnitine O-acetyltransferase; CAT; CAT1; CAT-1; Crat; Ecotropic retroviral leukemia receptor homolog; ecotropic retroviral receptor; ecotropic retrovirus receptor homolog; ERR; HCAT1; High affinity cationic amino acid transporter 1; REC1L; SLC7A1; solute carrier family 7 (cationic amino acid transporter, y+ system), member 1; solute carrier family 7 member 1; System Y+ basic amino acid transporter
Gene Symbols CRAT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 380-626 of human CRAT (NP_0007463).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12908, 1384, 311849
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.