Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAT1 (SLC7A1) Rat anti-Human, BUV661, Clone: CA2, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 756435
Description
CAT1 (SLC7A1) Monoclonal antibody specifically detects CAT1 (SLC7A1) in Human samples. It is validated for Flow cytometry
Specifications
CAT1 (SLC7A1) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
6541 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2b κ |
Flow Cytometry | |
CA2 | |
Brilliant Ultraviolet 661 | |
ATRC1; CAT-1; ERR; REC1L; SLC7A1; ecotropic retroviral leukemia receptor homolog | |
Human CAT1 Transfected Cells | |
50 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction