Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Catalase
SCI_ED

Catalog No. S25239 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
40 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25239 40 mg
S25239A 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25239

Oxidizing Enzyme

  • Powder
  • CAS Reg. 9001-05-2
  • Shelf life: 12 months
  • Recommended Storage: Refrigeration

This product is for educational use - no C of A is available.

Specifications

Concentration 20,000 U/g
Color White to Tan
Purity 100%
For Use With (Application) Laboratory chemicals
Content And Storage Refrigerated
Quantity 40 mg
Product Type Catalase
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.