Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Catalase Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299933FR
Description
Catalase Polyclonal specifically detects Catalase in Human samples. It is validated for ELISA.Specifications
Catalase | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
DyLight 680 | |
catalase, EC 1.11.1.6, MGC138422, MGC138424 | |
Produced in rabbits immunized with purified, recombinant Catalase (P04040; Ala2-Leu527). | |
0.1 mL | |
Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction | |
847 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction