Learn More
MP Biomedicals™ Catalase from Human Erythrocytes (E.C.1.11.1.6.)
Shop All MP Biomedicals, Inc ProductsDescription
Activity: approx. 160,000U/mg protein. Supplied as frozen solution of 1mg/mL in 1.5mM sodium phosphate, 20mM NaCl, pH 7.0 composed of four identical subunits. Unit definition: One unit is defined as the amount of enzyme that will decompose 1μmole of H2O2 per minute at 25°C, pH 7.0.
Ref.: Bonaventura, J., et al., Arch. Biochem. Biophysics, 150: 606 (1972).
Activity: approx. 160,000U/mg protein. Supplied as frozen solution of 1mg/mL in 1.5mM sodium phosphate, 20mM NaCl, pH 7.0 composed of four identical subunits. Unit definition: One unit is defined as the amount of enzyme that will decompose 1μmole of H2O2 per minute at 25°C, pH 7.0.
Ref.: Bonaventura, J., et al., Arch. Biochem. Biophysics, 150: 606 (1972).
Specifications
Specifications
| Concentration | 1 mg/mL |
| For Use With (Application) | Protecting cells against oxidative damage to proteins, lipids and nucleic acids |
| Storage Buffer | Frozen solution of 1mg/mL in 1.5mM Sodium Phosphate, 20mM NaCl, pH 7.0 |
| Content And Storage | -20°C |
| Source | Human erythrocyte |
| Quantity | 25 kU |
| Product Type | Catalase |
| Molecular Weight (g/mol) | 256000 |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.