Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Catalase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595349
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595349 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595349 Supplier Invitrogen™ Supplier No. PA595349
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Human Catalase shares 85.3% and 82.6% amino acid (aa) sequence identity with mouse and rat Catalase, respectively. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human A549 whole cell, human HL-60 whole cell, human THP-1 whole cell, human U937 whole cell, human HepG2 whole cell, human K562 whole cell, human HCCT tissue, human HCCP tissue, rat liver tissue, rat kidney tissue, rat lung tissue, rat RH35 whole cell, mouse liver tissue, mouse kidney tissue, mouse lung tissue. IHC: mouse liver tissue, rat liver tissue, human liver cancer tissue. ICC/IF: A431 cell. Flow: SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kDa in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Catalase
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene Cat
Gene Accession No. P04040, P04762, P24270
Gene Alias 2210418N07; Cas1; Cas-1; CAT; Cat01; catalase; catalase 1; Catl; CS1; Cs-1; CTA1; HGNC:1516; hypothetical protein LOC531682; MGC138422; MGC138424
Gene Symbols Cat
Host Species Rabbit
Immunogen E.coli-derived human Catalase recombinant protein (Position: E344-L527).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12359, 24248, 847
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.