Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Catalase Polyclonal Antibody, HRP
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA126784
Description
PA1-26784 is expected to cross react with all mammals and most aerobic microorganisms due to sequence homology.
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kDa in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara's disease.
Specifications
Catalase | |
Polyclonal | |
HRP | |
Cat | |
2210418N07; Cas1; Cas-1; CAT; Cat01; catalase; catalase 1; Catl; CS1; Cs-1; CTA1; HGNC:1516; hypothetical protein LOC531682; MGC138422; MGC138424 | |
Rabbit | |
IgG fraction | |
RUO | |
531682 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
10 mg/mL | |
0.02M potassium phosphate with 10mg/mL BSA, 0.15M NaCl and 0.01% gentamicin sulfate; pH 7.2 | |
P00432 | |
Cat | |
Catalase from bovine liver. | |
1 mg | |
Primary | |
Bovine | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction