Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catechol, 99%

Catalog No. aaa1016430
Click to view available options
Quantity:
250 g
5000 g

CAS: 120-80-9 | C6H6O2 | 110.11 g/mol

Catechol is used as a developer for photographic films, dyes and an intermediate for antioxidants in rubber and lubricating oils. It is used in polymerization inhibitors and in pharmaceuticals. It is a building block in organic synthesis and involved in the preparation of flavors and fragrances. It is used as a replacement for sandalwood oil, which is prepared from catechol through guaiacol and camphor. Further, it is a precursor for the preparation of vanillin from guaiacol, which is obtained by methylation reaction with catechol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Catechol is used as a developer for photographic films, dyes and an intermediate for antioxidants in rubber and lubricating oils. It is used in polymerization inhibitors and in pharmaceuticals. It is a building block in organic synthesis and involved in the preparation of flavors and fragrances. It is used as a replacement for sandalwood oil, which is prepared from catechol through guaiacol and camphor. Further, it is a precursor for the preparation of vanillin from guaiacol, which is obtained by methylation reaction with catechol.

Solubility
Soluble in water, pyridine, chloroform, benzene, carbon tetrachloride, ether and acetate.

Notes
Air and light sensitive. Incompatible with oxidizing agents, aluminum, bases, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-80-9
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00002188
InChI Key YCIMNLLNPGFGHC-UHFFFAOYSA-N
Synonym pyrocatechol, catechol, 1,2-dihydroxybenzene, 1,2-benzenediol, pyrocatechin, 2-hydroxyphenol, o-benzenediol, pyrocatechine, o-dihydroxybenzene, o-dioxybenzene
PubChem CID 289
ChEBI CHEBI:18135
IUPAC Name benzene-1,2-diol
SMILES OC1=CC=CC=C1O

Specifications

Melting Point 104°C to 107°C
Density 1.370
Boiling Point 245°C to 246°C
Flash Point 127°C (260°F)
Assay Percent Range 99%
Quantity 250 g
UN Number UN2811
Beilstein 471401
Sensitivity Air and light sensitive
Merck Index 14,7999
Solubility Information Soluble in water,pyridine,chloroform,benzene,carbon tetrachloride,ether and acetate.
Formula Weight 110.11
Percent Purity 99%
Chemical Name or Material Catechol
Show More Show Less
Hazard Category H301+H311-H315-H317-H318-H332-H341-H350-H500
Hazard Statement GHS H Statement
H301-H311-H318-H341-H351-H315
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 204-427-5
RTECSNumber UX1050000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.