Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Catenin alpha 1 Antibody (8B6C1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | Catenin alpha 1 |
---|---|
Clone | 8B6C1 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61722
![]() |
Novus Biologicals
NBP261722 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
Catenin alpha 1 Monoclonal specifically detects Catenin alpha 1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Catenin alpha 1 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Alpha E-catenin, alpha-catenin, alpha-E-catenin, alphaE-catenin, Cadherin-associated protein, CAP102, catenin (cadherin-associated protein), alpha 1 (102kD), catenin (cadherin-associated protein), alpha 1, 102kDa, catenin alpha-1, FLJ36832, FLJ52416, Renal carcinoma antigen NY-REN-13 | |
CTNNA1 | |
IgG1 | |
Protein G purified |
8B6C1 | |
Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
Monoclonal | |
Purified | |
Cancer, Cellular Markers, Core ESC Like Genes, Cytoskeleton Markers, Phospho Specific, Stem Cell Markers | |
1495 | |
Purified recombinant fragment of human CTNNA1 (AA: 371-574) expressed in E. Coli. | |
Primary | |
100 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title