Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-Catenin alpha-1 (Ser655, Thr658) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567515
Description
Predicted to react with Mouse samples.
Associates with the cytoplasmic domain of a variety of cadherins. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties. Can associate with both E- and N-cadherins. Originally believed to be a stable component of E-cadherin/catenin adhesion complexes and to mediate the linkage of cadherins to the actin cytoskeleton at adherens junctions. In contrast, cortical actin was found to be much more dynamic than E-cadherin/catenin complexes and CTNNA1 was shown not to bind to F-actin when assembled in the complex suggesting a different linkage between actin and adherence junctions components. The homodimeric form may regulate actin filament assembly and inhibit actin branching by competing with the Arp2/3 complex for binding to actin filaments. May play a crucial role in cell differentiation.
Specifications
Phospho-Catenin alpha-1 (Ser655, Thr658) | |
Polyclonal | |
Unconjugated | |
CTNNA1 | |
[a]E-catenin; 102 kDa cadherin-associated protein; 2010010M04Rik; AA517462; AI988031; alpha E catenin; alpha E-catenin; alpha(E)-catenin; alpha-E-catenin; cadherin associated protein; Cadherin-associated protein; CAP102; catenin (cadherin associated protein), alpha 1; catenin (cadherin-associated protein), alpha; catenin (cadherin-associated protein), alpha 1; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin alpha 1; catenin alpha 1 S homeolog; catenin alpha-1; catenin, alpha 1; Catna1; cell-adhesion protein alphaE catenin; ctnna; ctnna1; ctnna1.S; MDPT2; Renal carcinoma antigen NY-REN-13; XELAEV_18020018mg | |
Rabbit | |
Affinity chromatography | |
RUO | |
12385, 1495 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P26231, P35221 | |
CTNNA1 | |
Synthesized peptide derived from human alpha Catenin around the phosphorylation site of Ser655/Thr658. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction