Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Catenin alpha-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590334
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590334 Supplier Invitrogen™ Supplier No. PA590334
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TNEQDLANRF KEFGKEMVKL NYVAARRQQE LKDPHCRDEM AAARGALKKN ATMLYTASQA FLRHPDVAAT RANRDYVFKQ VQEAIAGISN AAQATSPTDE AKGHTGIGEL AAALNEFDNK IILD Positive Samples: Mouse brain, Rat brain; Cellular Location: Cell junction, Cell membrane, Cell projection, Cytoplasm, Cytoplasmic side, Peripheral membrane protein, adherens junction, axon, cytoskeleton.

Associates with the cytoplasmic domain of a variety of cadherins. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties. Can associate with both E- and N-cadherins. Originally believed to be a stable component of E-cadherin/catenin adhesion complexes and to mediate the linkage of cadherins to the actin cytoskeleton at adherens junctions. In contrast, cortical actin was found to be much more dynamic than E-cadherin/catenin complexes and CTNNA1 was shown not to bind to F-actin when assembled in the complex suggesting a different linkage between actin and adherence junctions components. The homodimeric form may regulate actin filament assembly and inhibit actin branching by competing with the Arp2/3 complex for binding to actin filaments. May play a crucial role in cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Catenin alpha-1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CTNNA1
Gene Accession No. P26231, P35221
Gene Alias [a]E-catenin; 102 kDa cadherin-associated protein; 2010010M04Rik; AA517462; AI988031; alpha E catenin; alpha E-catenin; alpha(E)-catenin; alpha-E-catenin; cadherin associated protein; Cadherin-associated protein; CAP102; catenin (cadherin associated protein), alpha 1; catenin (cadherin-associated protein), alpha; catenin (cadherin-associated protein), alpha 1; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin alpha 1; catenin alpha 1 S homeolog; catenin alpha-1; catenin, alpha 1; Catna1; cell-adhesion protein alphaE catenin; ctnna; ctnna1; ctnna1.S; MDPT2; Renal carcinoma antigen NY-REN-13; XELAEV_18020018mg
Gene Symbols CTNNA1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 167-290 of human CTNNA2 (NP_0012695261).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12385, 1495, 307505
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.