Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catenin delta-1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987655
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7655 Supplier Bioss Supplier No. BS7000R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.

Specifications

Antigen Catenin delta-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CTNND1
Gene Accession No. O60716
Gene Alias AA409437; AU019353; Cadherin-associated Src substrate; CAS; catenin (cadherin associated protein), delta 1; catenin (cadherin-associated protein), delta 1; catenin delta 1; Catenin delta-1; catenin src; Catns; CTNND; CTNND1; Kiaa0384; mKIAA0384; P120; p120 catenin; P120 p120; P120 P120CTN; p120(cas); p120(ctn); P120CAS; p120-catenin; P120CTN; RCJMB04_21j12
Gene Symbols CTNND1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Catenin delta-1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1500
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.