Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Catenin delta-2 Recombinant Rabbit Monoclonal Antibody (HL1629)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547114
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Armadillo-like proteins are characterized by a series of armadillo repeats, first defined in the Drosophila 'armadillo' gene product, that are typically 42 to 45 amino acids long. These proteins can be divided into subfamilies based on the number of repeats, their overall sequence similarity, and the dispersion of the repeats throughout their sequences. Members of the p120(ctn)/plakophilin subfamily of Armadillo-like proteins, including CTNND1, Catenin delta-2, PKP1, PKP2, PKP4, and ARVCF, have 10 armadillo repeats each (Bonne et al., 1998 [PubMed 9721216]).
Specifications
Catenin delta-2 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O35116, O35927, Q9UQB3 | |
CTNND2 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human delta 2 Catenin. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry | |
HL1629 | |
Unconjugated | |
CTNND2 | |
catenin (cadherin associated protein), delta 2; catenin (cadherin-associated protein), delta 2; catenin (cadherin-associated protein), delta 2 (neural plakophilin-related arm-repeat protein); catenin delta 2; catenin delta-2; catenin delta-2; LOW QUALITY PROTEIN: catenin delta-2; Catnd2; Ctnnd2; Delta-catenin; GT24; Neural plakophilin-related ARM-repeat protein; neurojugin; Neurojungin; Nprap; T-cell delta-catenin | |
Rabbit | |
Protein A | |
RUO | |
114028, 1501, 18163 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction