Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catenin delta-2 Mouse anti-Human, Mouse, Unconjugated, Clone: 4H2.1, MilliporeSigma™

Catalog No. MABN233025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2330-25 25 μg
MABN2330-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN2330-25 Supplier MilliporeSigma Supplier No. MABN233025UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Catenin delta-2, clone 4H2.1 Antibody, Cat. No. MABN2330, is a highly specific mouse monoclonal antibody that targets Catenin delta-2 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Catenin delta-2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4H2.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Delta-catenin;GT24;Neural plakophilin-related ARM-repeat protein;NPRAP;Neurojungin
Gene Symbols CTNND2;NPRAP
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 145 amino acids from the C-terminal region of human Catenin delta-2.
Molecular Weight of Antigen ∽150 kDa observed; 132.66 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001323
Test Specificity Clone 4H2.1 specifically detects Catenin delta-2 in human and murine cells. It targets an epitope with in the C-tereminal region.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.