Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catenin Delta-2 Mouse anti-Rat, Human, Clone: 12B4.1, MilliporeSigma™

Catalog No. MABN2254MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2254MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2254MI Supplier MilliporeSigma Supplier No. MABN2254
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Catenin Delta-2 Antibody, clone 12B4 is a highly specific mouse monoclonal antibody that targets CTNND2 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Catenin Delta-2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 12B4.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CTNND2,NPRAP
Host Species Mouse
Immunogen GST/His-tagged recombinat fragment corresponding to 104 amino acids from the C-terminal half of human CTNND2.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001323
Test Specificity Clone 12B4.1 specifically detects CTNND2 in rat brain microsomal preparations. This clone targets a sequeence within 104 amino acids in the C-terminal half of CTNND2.
Target Species Rat, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.