Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cathelicidin/FALL-39/LL-37, Mouse, Unlabeled, Clone: 8C10.1,

Catalog No. MABS1136MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1136MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1136MI Supplier MilliporeSigma™ Supplier No. MABS1136
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cathelicidin antimicrobial peptide (UniProt P49913; also known as 18kDa cationic antimicrobial protein, CAP-18, hCAP-18) is encoded by the CAMP (also known as CAP18, FALL39, LL37) gene (ORF HSD26; Gene ID 820) in human. The human cathelicidin antimicrobial protein 18 (hCAP-18) is initially translated as a precursor molecule with a signal peptide (a.a. 1-30) and a propeptide (a.a. 31-131) sequence, the removal of which yields the mature hCAP-18 (a.a. a.a. 132-173). During the inflammatory response, hCAP-18 is proteolytically cleaved into the FALL-39 (a.a 132-170), which is further processed to yield the antimicrobial protein LL-37 (CAP-37; a.a. 134-170). Neutrophils store preformed hCAP-18 within specific granules and release it into inflamed tissue as a result of granule exocytosis. Once released, hCAP-18 is activated by proteinase-3-mediated cleavage, a component of neutrophil primary granules. In addition to its microbicidal and LPS-sequestering activity, LL-37 is also involved in the regulation of innate and humoral immune responses, wound healing, and tissue repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathelicidin/FALL-39/LL-37
Applications Immunohistochemistry
Classification Monoclonal
Clone 8C10.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene CAMP, CAP18, FALL39, HSD26, LL37
Gene Accession No. P49913
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to the C-terminal sequence of human Cathelicidin/FALL-39/LL-37.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_004336
Test Specificity Clone 8C10.1 targets a C-terminal epitope present in the prepro- (a.a. 1-173) and pro- (a.a. 31-173) forms of hCAP-18, as well as the processed antibacterial peptides FALL-39 (a.a 132-170) and LL-37 (a.a. 134-170), but not the preprodomain (a.a. 1-131) or prodomain (a.a. 31-131) fragment.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.