Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin 3 Antibody (RM0156-2K13), DyLight 650, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212422C
Description
Cathepsin 3 Monoclonal specifically detects Cathepsin 3 in Mouse samples. It is validated for Western Blot.Specifications
Cathepsin 3 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Cts3 | |
Mouse recombinant Cathepsin 3 | |
0.1 mL | |
117066 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0156-2K13 | |
Western Blot | |
1600000I23Rik | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction